2-Ethynyl-4-fluorobenzonitrile 95%

Supplier: Apollo Scientific

PC502249-1G PC502249-250MG
APOSPC502249-1GEA 609 EUR
APOSPC502249-1G APOSPC502249-250MG
2-Ethynyl-4-fluorobenzonitrile 95%
2-Ethynyl-4-fluorobenzonitrile
Formula: C₉H₄FN
MW: 145.133 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11656151
CAS Number: 1208076-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR