2-Bromo-1-(3-bromo-5-chloro-2-fluorophenyl)ethanone
Supplier: Apollo Scientific
Synonyms: 3-Bromo-5-chloro-2-fluorophenacyl bromide
PC50232-1G PC50232-250MG
APOSPC50232-1GEA 493 EUR
APOSPC50232-1G APOSPC50232-250MG
2-Bromo-1-(3-bromo-5-chloro-2-fluorophenyl)ethanone
2-Bromo-1-(3-bromo-5-chloro-2-fluorophenyl)ethanone
Formula: C₈H₄Br₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD28733989
CAS Number: 1804382-39-5
UN: 3261
ADR: 8,III