3-Bromo-2,4,5-trifluorobenzonitrile 95%
Supplier: Apollo Scientific
Danger
PC502350-1G PC502350-250MG
APOSPC502350-1GEA 522 EUR
APOSPC502350-1G APOSPC502350-250MG
3-Bromo-2,4,5-trifluorobenzonitrile 95%
3-Bromo-2,4,5-trifluorobenzonitrile
Formula: C₇HBrF₃N
MW: 235.989 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756734
CAS Number: 104222-41-5
UN: 3439
ADR: 6.1 III