2-Nitro-5-(trifluoromethoxy)benzonitrile
Supplier: Apollo Scientific
PC502663-1G
PC502663-5G
APOSPC502663-1GEA
168
EUR
APOSPC502663-1G
APOSPC502663-5G
2-Nitro-5-(trifluoromethoxy)benzonitrile
2-Nitro-5-(trifluoromethoxy)benzonitrile
Formula:
C₈H₃F₃N₂O₃ MW: 232.116 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835205 CAS Number: 1003708-58-4 |