Methyl 3-chloro-5-fluorobenzoate
Supplier: Apollo Scientific
Synonyms: 3-Chloro-5-fluorobenzoic acid methyl ester
PC502675-1G PC502675-5G
APOSPC502675-1GEA 75.4 EUR
APOSPC502675-1G APOSPC502675-5G
Methyl 3-chloro-5-fluorobenzoate
Methyl 3-chloro-5-fluorobenzoate
Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204128
CAS Number: 327056-75-7