2-Bromo-5-(trifluoromethoxy)benzonitrile 95%
Supplier: Apollo Scientific
PC502682-1G PC502682-250MG PC502682-5G
APOSPC502682-1GEA 130 EUR
APOSPC502682-1G APOSPC502682-250MG APOSPC502682-5G
2-Bromo-5-(trifluoromethoxy)benzonitrile 95%
2-Bromo-5-(trifluoromethoxy)benzonitrile
Formula: C₈H₃BrF₃NO
MW: 266.015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28746664
CAS Number: 1804402-93-4
UN: 3276
ADR: 6.1,III