Methyl 3-bromo-4-(trifluoromethoxy)benzoate
Supplier: Apollo Scientific
PC502689-1G PC502689-5G
APOSPC502689-1GEA 40.6 EUR
APOSPC502689-1G APOSPC502689-5G
Methyl 3-bromo-4-(trifluoromethoxy)benzoate
Methyl 3-bromo-4-(trifluoromethoxy)benzoate
Formula: C₉H₆BrF₃O₃
MW: 299.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110912
CAS Number: 1131594-45-0