3-Bromo-2-fluorophenylisothiocyanate
Supplier: Apollo Scientific
Danger
PC503005-1G PC503005-25G PC503005-5G
APOSPC503005-1GEA 82.8 EUR
APOSPC503005-1G APOSPC503005-25G APOSPC503005-5G
3-Bromo-2-fluorophenylisothiocyanate
3-Bromo-2-fluorophenylisothiocyanate
Formula: C₇H₃BrFNS
MW: 232.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26686203
CAS Number: 1517289-83-6
UN: 2922
ADR: 8+6.1,III