3-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
Supplier: Apollo Scientific
Danger
PC503007-1G PC503007-25G PC503007-5G PC503007-100G
APOSPC503007-1GEA 87 EUR
APOSPC503007-1G APOSPC503007-25G APOSPC503007-5G APOSPC503007-100G
3-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
3-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
Formula: C₈H₃BrF₃NOS
MW: 298.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200430
CAS Number: 1379677-28-7
UN: 2922
ADR: 8+6.1,III