4-Bromo-3-fluorostyrene 95%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Bromo-3-fluorostyrene
PC50320-1G PC50320-250MG PC50320-5G
APOSPC50320-1GEA 516 EUR
APOSPC50320-1G APOSPC50320-250MG APOSPC50320-5G
4-Bromo-3-fluorostyrene 95%
4-Bromo-3-fluorostyrene
Formula: C₈H₆BrF
MW: 201.036 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24108136
CAS Number: 916814-07-8