2-Ethynyl-5-fluorobenzonitrile 95%
Supplier: Apollo Scientific
PC50558-1G PC50558-250MG
APOSPC50558-1GEA 609 EUR
APOSPC50558-1G APOSPC50558-250MG
2-Ethynyl-5-fluorobenzonitrile 95%
2-Ethynyl-5-fluorobenzonitrile
Formula: C₉H₄FN
MW: 145.133 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18205104
CAS Number: 1378823-86-9