3-Ethynyl-4-fluorobenzonitrile 95%
Supplier: Apollo Scientific
PC50561-1G
PC50561-250MG
APOSPC50561-1GEA
609
EUR
APOSPC50561-1G
APOSPC50561-250MG
3-Ethynyl-4-fluorobenzonitrile 95%
3-Ethynyl-4-fluorobenzonitrile
Formula:
C₉H₄FN MW: 145.133 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11618087 CAS Number: 133960-64-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...