5-Bromo-8-(trifluoromethyl)quinolin-4(1H)-one
Supplier: Apollo Scientific
Danger
PC508119-1G PC508119-250MG
APOSPC508119-1GEA 241 EUR
APOSPC508119-1G APOSPC508119-250MG
5-Bromo-8-(trifluoromethyl)quinolin-4(1H)-one
5-Bromo-8-(trifluoromethyl)quinolin-4(1H)-one
Formula: C₁₀H₅BrF₃NO
MW: 292.052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31745740
CAS Number: 1065092-40-1
UN: 2811
ADR: 6.1,III