4,6-Dichloro-8-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
Warning
PC508132-1G PC508132-250MG
APOSPC508132-1GEA 281 EUR
APOSPC508132-1G APOSPC508132-250MG
4,6-Dichloro-8-(trifluoromethyl)quinoline
4,6-Dichloro-8-(trifluoromethyl)quinoline
Formula: C₁₀H₄Cl₂F₃N
MW: 266.047 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28010395
CAS Number: 1065093-28-8