1-(6-Bromo-2,3,4-trifluorophenyl)ethanone 95%
Supplier: Apollo Scientific
PC50948-1G
PC50948-250MG
APOSPC50948-1GEA
609
EUR
APOSPC50948-1G
APOSPC50948-250MG
1-(6-Bromo-2,3,4-trifluorophenyl)ethanone 95%
1-(6-Bromo-2,3,4-trifluorophenyl)ethanone
Formula:
C₈H₄BrF₃O MW: 253.016 g/mol Storage Temperature: Ambient |
CAS Number:
1509119-02-1 |