1-(3-Bromo-5-chloro-2-fluorophenyl)ethanone 95%
Supplier: Apollo Scientific
PC50972-1G PC50972-250MG PC50972-5G
APOSPC50972-1GEA 493 EUR
APOSPC50972-1G APOSPC50972-250MG APOSPC50972-5G
1-(3-Bromo-5-chloro-2-fluorophenyl)ethanone 95%
1-(3-Bromo-5-chloro-2-fluorophenyl)ethanone
Formula: C₈H₅BrClFO
MW: 251.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32693996
CAS Number: 2092624-18-3