1-Bromo-2-(2,2-diethoxyethoxy)-4-fluorobenzene
Supplier: Apollo Scientific
PC510010-1G PC510010-5G
APOSPC510010-1GEA 194 EUR
APOSPC510010-1G APOSPC510010-5G
1-Bromo-2-(2,2-diethoxyethoxy)-4-fluorobenzene
1-Bromo-2-(2,2-diethoxyethoxy)-4-fluorobenzene
Formula: C₁₂H₁₆BrFO₃
MW: 307.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191631
CAS Number: 253429-30-0