Methyl 4-Fluoro-2-(trifluoroacetamido)benzoate
Supplier: Apollo Scientific
PC510012-25G PC510012-5G
APOSPC510012-25GEA 549 EUR
APOSPC510012-25G APOSPC510012-5G
Methyl 4-Fluoro-2-(trifluoroacetamido)benzoate
Methyl 4-Fluoro-2-(trifluoroacetamido)benzoate
Formula: C₁₀H₇F₄NO₃
MW: 265.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14585008
CAS Number: 404010-71-5