[6-(2,4-Difluorophenyl)-2-pyridyl]methanol
Supplier: Apollo Scientific
PC510060-1G PC510060-5G
APOSPC510060-1GEA 499 EUR
APOSPC510060-1G APOSPC510060-5G
[6-(2,4-Difluorophenyl)-2-pyridyl]methanol
[6-(2,4-Difluorophenyl)-2-pyridyl]methanol
Formula: C₁₂H₉F₂NO
MW: 221.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410125
CAS Number: 887981-41-1