[6-[2-(Trifluoromethyl)phenyl]3-pyridyl]methanol
Supplier: Apollo Scientific
PC510092-1G PC510092-5G
APOSPC510092-1GEA 499 EUR
APOSPC510092-1G APOSPC510092-5G
[6-[2-(Trifluoromethyl)phenyl]3-pyridyl]methanol
[6-[2-(Trifluoromethyl)phenyl]3-pyridyl]methanol
Formula: C₁₃H₁₀F₃NO
MW: 253.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410446
CAS Number: 220455-42-5