2-Bromo-3-fluoro-6-nitroaniline 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Amino-3-bromo-4-fluoronitrobenzene
PC51010-1G PC51010-5G
APOSPC51010-1GEA 34.8 EUR
APOSPC51010-1G APOSPC51010-5G
2-Bromo-3-fluoro-6-nitroaniline 97%
2-Bromo-3-fluoro-6-nitroaniline
Formula: C₆H₄BrFN₂O₂
MW: 235.011 g/mol
Boiling Pt: 315 °C
Melting Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18071077
CAS Number: 1218764-80-7
UN: 2811
ADR: 6.1 III