3-Fluoro-5-(hydroxymethyl)-2-methylpyridine
Supplier: Apollo Scientific
PC510205-1G
PC510205-250MG
APOSPC510205-1GEA
1730
EUR
APOSPC510205-1G
APOSPC510205-250MG
3-Fluoro-5-(hydroxymethyl)-2-methylpyridine
3-Fluoro-5-(hydroxymethyl)-2-methylpyridine
Formula:
C₇H₈FNO MW: 141.143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19440436 CAS Number: 917835-69-9 |