(S)-3-(3-Fluorophenyl)-2-hydroxypropionic acid
Supplier: Apollo Scientific
PC510276-5G PC510276-1G
APOSPC510276-5GEA 1330 EUR
APOSPC510276-5G APOSPC510276-1G
(S)-3-(3-Fluorophenyl)-2-hydroxypropionic acid
(S)-3-(3-Fluorophenyl)-2-hydroxypropionic acid
Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977372
CAS Number: 159415-52-8