Methyl 3-bromo-5-fluoro-2-methylbenzoate
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-5-fluoro-2-methyl-benzoic acid methyl ester
PC510281-1G PC510281-250MG
APOSPC510281-1GEA 188 EUR
APOSPC510281-1G APOSPC510281-250MG
Methyl 3-bromo-5-fluoro-2-methylbenzoate
Methyl 3-bromo-5-fluoro-2-methylbenzoate
Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440363
CAS Number: 1187318-53-1