7-Fluoro-2-hydroxynaphthalene-1,4-dione
Supplier: Apollo Scientific
PC510289-1G PC510289-250MG
APOSPC510289-1GEA 1620 EUR
APOSPC510289-1G APOSPC510289-250MG
7-Fluoro-2-hydroxynaphthalene-1,4-dione
7-Fluoro-2-hydroxynaphthalene-1,4-dione
Formula: C₁₀H₅FO₃
MW: 192.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14706170
CAS Number: 58472-36-9