1-[(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methyl]imidazole
Supplier: Apollo Scientific
Warning
Synonyms: 1-((2-Fluorophenyl)(4-fluorophenyl)(phenyl)methyl)-1H-imidazole
PC510299-1G PC510299-250MG PC510299-5G
APOSPC510299-1GEA 591 EUR
APOSPC510299-1G APOSPC510299-250MG APOSPC510299-5G
1-[(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methyl]imidazole
1-[(2-Fluorophenyl)(4-fluorophenyl)(phenyl)methyl]imidazole
Formula: C₂₂H₁₆F₂N₂
MW: 346.373 g/mol
Melting Pt: 161 °C
Storage Temperature: Ambient
MDL Number: MFCD00865591
CAS Number: 119006-77-8