1,2,2-Tribromo-1H,1H-tridecafluorooctane
Supplier: Apollo Scientific
Warning
Synonyms: 7,7,8-Tribromo-1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorooctane, 1-(Perfluorohex-1-yl)-1,1,2-tribromoethane, 2,2-Dibromo-2-(perfluorohex-1-yl)ethyl bromide
PC51060-1G PC51060-25G PC51060-5G
APOSPC51060-1GEA 73 EUR
APOSPC51060-1G APOSPC51060-25G APOSPC51060-5G
1,2,2-Tribromo-1H,1H-tridecafluorooctane
1,2,2-Tribromo-1H,1H-tridecafluorooctane
Formula: C₈H₂Br₃F₁₃
MW: 584.793 g/mol
Boiling Pt: 116 °C (1 mm Hg)
Melting Pt: 28…29 °C
Storage Temperature: Ambient
MDL Number: MFCD18641926
CAS Number: 59665-25-7