1,2,3,4,5,-Pentafluoro-6-[(1E)-prop-1-en-1-yl]benzene
Supplier: Apollo Scientific
Warning
Synonyms: trans-1-(Perfluorophenyl)prop-1-ene, trans-1-(Pentafluorophenyl)prop-1-ene
PC51070-1G PC51070-5G
APOSPC51070-1GEA 228 EUR
APOSPC51070-1G APOSPC51070-5G
1,2,3,4,5,-Pentafluoro-6-[(1E)-prop-1-en-1-yl]benzene
1,2,3,4,5,-Pentafluoro-6-[(1E)-prop-1-en-1-yl]benzene
Formula: C₉H₅F₅
MW: 208.128 g/mol
Boiling Pt: 86 °C
Density: 1.374 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD19442035
CAS Number: 4683-67-4
UN: 1993
ADR: 3,III