1,1-Difluoro-2-phenylpropan-2-ol 97%
Supplier: Apollo Scientific
PC51087-1G PC51087-5G
APOSPC51087-1GEA 429 EUR
APOSPC51087-1G APOSPC51087-5G
1,1-Difluoro-2-phenylpropan-2-ol 97%
1,1-Difluoro-2-phenylpropan-2-ol
Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Melting Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD08447367
CAS Number: 432-88-2