Methyl 4-fluoro-3-iodobenzoate 97%
Supplier: Apollo Scientific
Warning
PC51092-1G PC51092-5G PC51092-25G
APOSPC51092-1GEA 21.9 EUR
APOSPC51092-1G APOSPC51092-5G APOSPC51092-25G
Methyl 4-fluoro-3-iodobenzoate 97%
Methyl 4-fluoro-3-iodobenzoate
Formula: C₈H₆FIO₂
MW: 280.035 g/mol
Boiling Pt: 287…288 °C
Melting Pt: 42…43 °C
Density: 1.823 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD11870177
CAS Number: 1121586-29-5