4,5-Diamino-2-(trifluoromethyl)benzonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Cyano-4,5-diaminobenzotrifluoride, 4-Cyano-5-(trifluoromethyl)benzene-1,2-diamine
PC51113-1G PC51113-5G
APOSPC51113-1GEA 96.3 EUR
APOSPC51113-1G APOSPC51113-5G
4,5-Diamino-2-(trifluoromethyl)benzonitrile 98%
4,5-Diamino-2-(trifluoromethyl)benzonitrile
Formula: C₈H₆F₃N₃
Melting Pt: 162…163 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD18074368
CAS Number: 882978-62-3
UN: 3439
ADR: 6.1,III