2,6-Difluoro-3-hydroxypyridine 96%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Difluoro-3-hydroxy-pyridine, 2,6-Difluoropyridin-3-ol
PC51115-1G PC51115-250MG PC51115-5G
APOSPC51115-1GEA 156 EUR
APOSPC51115-1G APOSPC51115-250MG APOSPC51115-5G
2,6-Difluoro-3-hydroxypyridine 96%
2,6-Difluoro-3-hydroxypyridine
Formula: C₅H₃F₂NO
Melting Pt: 124…125 °C
Storage Temperature: Ambient
MDL Number: MFCD11044351
CAS Number: 209328-85-8