(2S)-1,1,1-Trifluoropropan-2-ol 97%
Supplier: Apollo Scientific
Warning
PC51130-1G PC51130-5G
APOSPC51130-1GEA 161 EUR
APOSPC51130-1G APOSPC51130-5G
(2S)-1,1,1-Trifluoropropan-2-ol 97%
Formula: C₃H₅F₃O
Boiling Pt: 77…78 °C (754 mm Hg)
Melting Pt: –52 °C
Density: 1.2799 g/cm³ (25 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD06797331
CAS Number: 3539-97-7
UN: 1987
ADR: 3,II