2,6-Difluoro-3-methoxybenzamide
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Aminocarbonyl)-2,4-difluoroanisole, 3-Carbamoyl-2,4-difluoroanisole
PC51140-10G PC51140-5G
APOSPC51140-10GEA 488 EUR
APOSPC51140-10G APOSPC51140-5G
2,6-Difluoro-3-methoxybenzamide
2,6-Difluoro-3-methoxybenzamide
Formula: C₈H₇F₂NO₂
MW: 187.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115900
CAS Number: 886498-26-6