4-Fluoro-α-methylbenzyl bromide
Supplier: Apollo Scientific
Warning
Synonyms: 1-(4-Fluorophenyl)ethyl bromide, 1-Bromo-1-(4-fluorophenyl)ethane, 1-(1-Bromoethyl)-4-fluorobenzene
PC51144-10G PC51144-5G
APOSPC51144-10GEA 102 EUR
APOSPC51144-10G APOSPC51144-5G
4-Fluoro-α-methylbenzyl bromide
4-Fluoro-α-methylbenzyl bromide
Formula: C₈H₈BrF
MW: 203.052 g/mol
Boiling Pt: 75…78 °C (1 mm Hg)
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11180274
CAS Number: 65130-46-3
UN: 2810
ADR: 6.1 III