3-Bromo-4-hydroxy-5-nitrobenzotrifluoride
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-6-nitro-4-(trifluoromethyl)phenol, 3-Bromo-2-hydroxy-5-(trifluoromethyl)nitrobenzene
PC51156-1G PC51156-5G
APOSPC51156-1GEA 306 EUR
APOSPC51156-1G APOSPC51156-5G
3-Bromo-4-hydroxy-5-nitrobenzotrifluoride
3-Bromo-4-hydroxy-5-nitrobenzotrifluoride
Formula: C₇H₃BrF₃NO₃
MW: 286.003 g/mol
Boiling Pt: 64…68 °C (1 mm Hg)
Melting Pt: 21…23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01569845
CAS Number: 206759-48-0