2-Chloro-6-(trifluoromethyl)quinoxaline
Supplier: Apollo Scientific
Warning
Synonyms: 2-Chloro-6-(trifluoromethyl)-1,4-benzodiazine
PC51163-1G PC51163-250MG
APOSPC51163-1GEA 219 EUR
APOSPC51163-1G APOSPC51163-250MG
2-Chloro-6-(trifluoromethyl)quinoxaline
2-Chloro-6-(trifluoromethyl)quinoxaline
Formula: C₉H₄ClF₃N₂
MW: 232.59 g/mol
Boiling Pt: 262.2 °C
Melting Pt: 117…120 °C
Flash Pt: 125.6 °C
Storage Temperature: Ambient
MDL Number: MFCD05669533
CAS Number: 41213-32-5