3-[4-(Trifluoromethyl)phenyl]propan-1-ol
Supplier: Apollo Scientific
Warning
Synonyms: 3-(4-(Trifluoromethyl)phenyl)propan-1-ol, 4-(3-Hydroxyprop-1-yl)benzotrifluoride
PC51191-1G PC51191-5G
APOSPC51191-1GEA 143 EUR
APOSPC51191-1G APOSPC51191-5G
3-[4-(Trifluoromethyl)phenyl]propan-1-ol
3-[4-(Trifluoromethyl)phenyl]propan-1-ol
Formula: C₁₀H₁₁F₃O
MW: 204.189 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09028744
CAS Number: 180635-74-9