2-(Difluoromethyl)-4-(hydroxymethyl)pyridine
Supplier: Apollo Scientific
Warning
Synonyms: (2-(Difluoromethyl)pyridin-4-yl)methanol, [2-(Difluoromethyl)pyridin-4-yl]methanol, 2-(Difluoromethyl)isonicotinyl alcohol
PC51207-1G PC51207-250MG
APOSPC51207-1GEA 578 EUR
APOSPC51207-1G APOSPC51207-250MG
2-(Difluoromethyl)-4-(hydroxymethyl)pyridine
2-(Difluoromethyl)-4-(hydroxymethyl)pyridine
Formula: C₇H₇F₂NO
MW: 159.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23701109
CAS Number: 1428532-80-2