4-Chloro-6-(trifluoromethyl)nicotinaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-5-formyl-2-(trifluoromethyl)pyridine, 4-Chloro-6-(trifluoromethyl)pyridine-3-carboxaldehyde
PC51209-1G PC51209-250MG
APOSPC51209-1GEA 687 EUR
APOSPC51209-1G APOSPC51209-250MG
4-Chloro-6-(trifluoromethyl)nicotinaldehyde
4-Chloro-6-(trifluoromethyl)nicotinaldehyde
Formula: C₇H₃ClF₃NO
MW: 209.553 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13188806
CAS Number: 1060807-48-8