3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
Supplier: Apollo Scientific
Warning
PC51362-1G PC51362-250MG
APOSPC51362-1GEA 133 EUR
APOSPC51362-1G APOSPC51362-250MG
3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
3-[3-Chloro-4-(trifluoromethoxy)phenyl]propanoic acid
Formula: C₁₀H₈ClF₃O₃
MW: 268.617 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393906
CAS Number: 1261873-29-3