2-Chloro-6-(trifluoromethyl)isonicotinaldehyde
Supplier: Apollo Scientific
PC51496-1G PC51496-250MG
APOSPC51496-1GEA 603 EUR
APOSPC51496-1G APOSPC51496-250MG
2-Chloro-6-(trifluoromethyl)isonicotinaldehyde
2-Chloro-6-(trifluoromethyl)isonicotinaldehyde
Formula: C₇H₃ClF₃NO
MW: 209.553 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188805
CAS Number: 1060807-47-7