2-(Trifluoromethylthio)benzonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Cyanophenyl trifluoromethyl sulphide
PC5179-10G PC5179-1G PC5179-5G
APOSPC5179-10GEA 980 EUR
APOSPC5179-10G APOSPC5179-1G APOSPC5179-5G
2-(Trifluoromethylthio)benzonitrile 98%
2-(Trifluoromethylthio)benzonitrile
Formula: C₈H₄F₃NS
Storage Temperature: Ambient
MDL Number: MFCD00236338
CAS Number: 37526-65-1