4-(2,2,2-Trifluoroethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-β,β,β-trifluorophenetole
PC5198-1G PC5198-5G
APOSPC5198-1GEA 26.1 EUR
APOSPC5198-1G APOSPC5198-5G
4-(2,2,2-Trifluoroethoxy)aniline 98%
4-(2,2,2-Trifluoroethoxy)aniline
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Melting Pt: 63…66 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077606
CAS Number: 57946-61-9
UN: 2811
ADR: 6.1 III