1-(1,1-Difluoroethyl)-4-nitrobenzene 97%
Supplier: Apollo Scientific
Warning
PC520501-1G PC520501-5G
APOSPC520501-1GEA 72.5 EUR
APOSPC520501-1G APOSPC520501-5G
1-(1,1-Difluoroethyl)-4-nitrobenzene 97%
1-(1,1-Difluoroethyl)-4-nitrobenzene
Formula: C₈H₇F₂NO₂
MW: 187.143 g/mol
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00034014
CAS Number: 32471-55-9