4,6-Dichloro-5-nitro-2-(trifluoromethyl)-pyrimidine 95%
Supplier: Apollo Scientific
PC520569-1G PC520569-5G
APOSPC520569-1GEA 280 EUR
APOSPC520569-1G APOSPC520569-5G
4,6-Dichloro-5-nitro-2-(trifluoromethyl)-pyrimidine 95%
4,6-Dichloro-5-nitro-2-(trifluoromethyl)-pyrimidine
Formula: C₅Cl₂F₃N₃O₂
MW: 261.974 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436609
CAS Number: 715-46-8