4-Bromo-1-fluoro-2-(2,2,2-trifluoroethyl)benzene 95%

Supplier: Apollo Scientific

APOSPC520617-5GEA 919 EUR
APOSPC520617-5G
4-Bromo-1-fluoro-2-(2,2,2-trifluoroethyl)benzene 95%
4-Bromo-1-fluoro-2-(2,2,2-trifluoroethyl)benzene
Formula: C₈H₅BrF₄
MW: 257,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11226694
CAS Number: 1186195-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR