2-Chloro-4-(1,1-difluoroethyl)pyridine 95%
Supplier: Apollo Scientific
PC520696-1G PC520696-5G
APOSPC520696-1GEA 268 EUR
APOSPC520696-1G APOSPC520696-5G
2-Chloro-4-(1,1-difluoroethyl)pyridine 95%
2-Chloro-4-(1,1-difluoroethyl)pyridine
Formula: C₇H₆ClF₂N
MW: 177.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14525494
CAS Number: 1204295-63-5