2-Hydroxy-3-fluoro-4-(trifluoromethyl)pyridine 97%
Supplier: Apollo Scientific
PC520732-1G PC520732-250MG
APOSPC520732-1GEA 663 EUR
APOSPC520732-1G APOSPC520732-250MG
2-Hydroxy-3-fluoro-4-(trifluoromethyl)pyridine 97%
2-Hydroxy-3-fluoro-4-(trifluoromethyl)pyridine
Formula: C₆H₃F₄NO
MW: 181.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611126
CAS Number: 1227594-89-9