2-Bromo-1-(difluoromethoxy)-3-fluorobenzene 95%
Supplier: Apollo Scientific
PC520739-1G PC520739-5G
APOSPC520739-1GEA 43.5 EUR
APOSPC520739-1G APOSPC520739-5G
2-Bromo-1-(difluoromethoxy)-3-fluorobenzene 95%
2-Bromo-1-(difluoromethoxy)-3-fluorobenzene
Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18390895
CAS Number: 1239492-22-8